Learn More
S100A9 Antibody (CL11191) [Janelia Fluor™ 549], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP3-07983JF549
Product Code. 30288050
  • 748.00€ / 0.10mL
Description
S100A9 Monoclonal antibody specifically detects S100A9 in Human samples. It is validated for Immunohistochemistry,Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
S100A9
Monoclonal
Janelia Fluor 549
CAGBMigration inhibitory factor-related protein 14, calgranulin B, calgranulin-B, Calprotectin L1H subunit, CFAGMRP-14,60B8AG, CGLB, L1AG, LIAG, MAC387, MIF, MRP14Leukocyte L1 complex heavy chain, NIF, P14, protein S100-A9, S100 calcium binding protein A9, S100 calcium binding protein A9 (calgranulin B), S100 calcium-binding protein A9, S100 calcium-binding protein A9 (calgranulin B)
This antibody was developed using a recombinant protein derived from P06702, with the exact immunogen sequence remaining proprietary.
0.1 mL
Cancer, Immunology, Innate Immunity, Signal Transduction, Tyrosine Kinases
6280
Store at 4⪚C in the dark.
IgG2a
Immunohistochemistry, Immunohistochemistry (Paraffin)
CL11191
50mM Sodium Borate