missing translation for 'onlineSavingsMsg'
Learn More
Learn More
S100A8 Antibody (2C5/4) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
214.00€ - 499.00€
Specifications
| Antigen | S100A8 |
|---|---|
| Clone | 2C5/4 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot : 1:100 - 1:2000, ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18634829
|
Novus Biologicals
NBP2-76650-0.025ml |
0.025 mL |
214.00€
0.03mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18362911
|
Novus Biologicals
NBP2-76650 |
0.1 mL |
499.00€
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
S100A8 Monoclonal specifically detects S100A8 in Human samples. It is validated for Western Blot, ELISA.Specifications
| S100A8 | |
| 1 mg/mL | |
| Monoclonal | |
| Mouse | |
| PBS with 0.02% sodium azide | |
| 6279 | |
| Recombinant human S100A8 | |
| Primary | |
| This antibody is specific for S100A8 both as an isolated protein and in combination with S100A9. |
| 2C5/4 | |
| Western Blot : 1:100 - 1:2000, ELISA | |
| Unconjugated | |
| Human | |
| 60B8AG, CAGACP-10, calgranulin A, calgranulin-A, Calprotectin L1L subunit, CFAGL1Ag, CGLA, Cystic fibrosis antigen, Leukocyte L1 complex light chain, MA387, MIF, Migration inhibitory factor-related protein 8, MRP-8, MRP8S100 calcium binding protein A8 (calgranulin A), NIF, P8, protein S100-A8, S100 calcium binding protein A8, S100 calcium-binding protein A8, S100 calcium-binding protein A8 (calgranulin A), Urinary stone protein band A | |
| S100A8 | |
| IgG1 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title