missing translation for 'onlineSavingsMsg'
Learn More
Learn More
S100A4 Rabbit anti-Human, Mouse, Clone: S100A4/2750R, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-07587-100ug
This item is not returnable.
View return policy
Description
S100A4 Monoclonal specifically detects S100A4 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| S100A4 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Western Blot 1 to 2 μg/mL, Flow Cytometry 1 to 2 μg/million cells, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1 to 2 μg/mL, Immunohistochemistry-Paraffin | |
| 18A2, Calvasculin, CAPLS100 calcium binding protein A4 (calcium protein, calvasculin, metastasin, fibroblast-specific protein-1,42A, FSP1, leukemia multidrug resistance associated protein, malignant transformation suppression 1, Metastasin, MTS1, murine placental homolog), P9KA, PEL98, Placental calcium-binding protein, Protein Mts1, protein S100-A4, S100 calcium binding protein A4, S100 calcium-binding protein A4, S100 calcium-binding protein A4 (calcium protein, calvasculin, metastasin, murine placental homolog) | |
| Recombinant fragment of human S100A4 protein (around aa1-200) (exact sequence is proprietary) | |
| 100 μg | |
| Cancer | |
| 6275 | |
| Store at 4°C. | |
| IgG |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| S100A4/2750R | |
| Unconjugated | |
| 10 mM PBS with 0.05% BSA | |
| Rabbit | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction