missing translation for 'onlineSavingsMsg'
Learn More
Learn More
S100A4, Goat anti-Human, Polyclonal Antibody, Abnova™
Goat Polyclonal Antibody
Brand: Abnova PAB7142.100ug
This item is not returnable.
View return policy
Description
Sequence: CNEFFEGFPDKQPSpecifications
| S100A4 | |
| Polyclonal | |
| Unconjugated | |
| In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide) | |
| 18A2/42A/CAPL/FSP1/MTS1/P9KA/PEL98 | |
| Goat | |
| 100 μg | |
| Primary | |
| Both variants (NP_002952.1; NP_062427.1) represent identical protein. | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| ELISA (1:128000) Western blot (0.03-0.1 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (2-3 ug/mL) The optimal working dilution should be determined by the end user. | |
| S100 calcium binding protein A4 | |
| S100A4 | |
| A synthetic peptide corresponding to human S100A4. | |
| RUO | |
| 6275 | |
| Human | |
| Antibody |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction