Learn More
S-Sulfo-L-cysteine Sodium Salt (~85%), TRC
Product Code. 30012518
Change view
Click to view available options
Quantity:
50 mg
100 mg
500 mg
Unit Size:
100mg
500mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30012518 100 mg 100mg
30011265 500 mg 500mg
30011062 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30012518 Supplier Toronto Research Chemicals Supplier No. TRCS789483100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 7381-67-1
Molecular Formula C3H6NO5S2Na
Molecular Weight (g/mol) 223.2
Synonym L-Cysteine, Hydrogen Sulfate (Ester), Monosodium Salt (6CI,8CI), L-Cysteine, Hydrogen Sulfate (Ester), Monosodium Salt (9CI);NSC 344479, S-Sulfocysteine Monosodium Salt, S-Sulfocysteine Sodium Salt, Sodium Cysteine-S-sulfonate
IUPAC Name sodium;(2R)-2-amino-1-hydroxy-1-oxo-3-sulfonatosulfanylpropane
SMILES [Na+].N[C@@H](CSS(=O)(=O)[O-])C(=O)O
Specifications
InChI Formula InChI=1S/C3H7NO5S2.Na/c4-2(3(5)6)1-10-11(7,8)9;/h2H,1,4H2,(H,5,6)(H,7,8,9);/q;+1/p-1/t2-;/m0./s1
Quantity 100 mg
Formula Weight 222.9585
Shipping Condition Room Temperature
Chemical Name or Material S-Sulfo-L-cysteine Sodium Salt