CAS RN 7381-67-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
S-Sulfo-L-cysteine Sodium Salt (~85%), TRC
CAS: 7381-67-1 Molecular Formula: C3H6NO5S2Na Molecular Weight (g/mol): 223.2 Synonym: L-Cysteine, Hydrogen Sulfate (Ester), Monosodium Salt (6CI,8CI),L-Cysteine, Hydrogen Sulfate (Ester), Monosodium Salt (9CI);NSC 344479,S-Sulfocysteine Monosodium Salt,S-Sulfocysteine Sodium Salt,Sodium Cysteine-S-sulfonate IUPAC Name: sodium;(2R)-2-amino-1-hydroxy-1-oxo-3-sulfonatosulfanylpropane SMILES: [Na+].N[C@@H](CSS(=O)(=O)[O-])C(=O)O
2
Tocris Bioscience™ S-Sulfo-L-cysteine sodium salt
Group I agonist