missing translation for 'onlineSavingsMsg'
Learn More

(S)-N-Desmethyl Citalopram Hydrochloride, TRC

Product Code. 17950845
Change view
Click to view available options
Quantity:
5 mg
50 mg
Unit Size:
50mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17950845 50 mg 50mg
17901785 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17950845 Supplier Toronto Research Chemicals Supplier No. TRCD29157150MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula C19H20ClFN2O
Molecular Weight (g/mol) 346.83
Synonym (1S)-1-(4-Fluorophenyl)-1,3-dihydro-1-[3-(methylamino)propyl]-5-isobenzo-furancarbonitrile Hydrochloride, (S)-1-(3-Methylaminopropyl)-1-(4-fluorophenyl)-1,3-dihydroisobenzofuran-5-carbonitrile Hydrochloride, (S)-(+)-N-Demethylcitalopram Hydrochloride;
SMILES Cl.CNCCC[C@]1(OCc2cc(ccc12)C#N)c3ccc(F)cc3

Specifications

Analyte or Component Names (S)-N-Desmethyl Citalopram Hydrochloride
Shipping Condition Room Temperature
InChI Formula InChI=1S/C19H19FN2O.ClH/c1-22-10-2-9-19(16-4-6-17(20)7-5-16)18-8-3-14(12-21)11-15(18)13-23-19;/h3-8,11,22H,2,9-10,13H2,1H3;1H/t19-;/m0./s1
Solution Type Neat
Industry Type Pharmaceutical
Quantity 50 mg
Percent Purity >95%
Chemical Name or Material (S)-N-Desmethyl Citalopram Hydrochloride
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.