missing translation for 'onlineSavingsMsg'
Learn More

(S)-(-)-1-(4-Chlorophenyl)ethylamine, ChiPros™ 97%, ee 98%

Product Code. 11359595
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11359595 25 g 25g
11339595 1 g 1g
11349595 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11359595 Supplier Thermo Scientific Alfa Aesar Supplier No. L19052.14

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 4187-56-8 | C8H10ClN | 155.625 g/mol

It is used as a substrate in palladium catalysts on alkaline-earth supports for racemization and dynamic kinetic resolution of benzylic amines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a substrate in palladium catalysts on alkaline-earth supports for racemization and dynamic kinetic resolution of benzylic amines.

Solubility
Soluble in water at 25°C 11 g/L.

Notes
Air sensitive. Store away from acids, air, acid chlorides, acid anhydrides and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4187-56-8
Molecular Formula C8H10ClN
Molecular Weight (g/mol) 155.625
MDL Number MFCD00671640
InChI Key PINPOEWMCLFRRB-LURJTMIESA-N
Synonym s-1-4-chlorophenyl ethylamine, s-1-4-chlorophenyl ethanamine, 1s-1-4-chlorophenyl ethanamine, s-4-chloro-alpha-methylbenzylamine, s---1-4-chlorophenyl ethylamine, 1s-1-4-chlorophenyl ethan-1-amine, pubchem15221, s-4-chloro-, a-methylbenzylamine, 1s-1-4-chlorophenyl ethylamine, s-1-4-chloro-phenyl ethylamine
PubChem CID 1550852
IUPAC Name (1S)-1-(4-chlorophenyl)ethanamine
SMILES CC(C1=CC=C(C=C1)Cl)N

Specifications

Density 1.11
Quantity 25 g
Odor Pungent
Refractive Index 1.5418
UN Number UN2922
Beilstein 2412316
Boiling Point 232°C
Solubility Information Soluble in water at 25°C 11g/L.
Formula Weight 155.63
Percent Purity 97%
Assay ee 98%
Chemical Name or Material (S)-(-)-1-(4-Chlorophenyl)ethylamine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.