accessibility menu, dialog, popup

UserName

CAS RN 4187-56-8

IUPAC Name: 1-(4-chlorophenyl)ethan-1-amine
Synonyms: 1-(4-chlorophenyl)ethanamine
Molecular Weight (g/mol): 155.63
Molecular Formula: C8H10ClN
InChi Key: PINPOEWMCLFRRB-UHFFFAOYNA-N
SMILES: CC(N)C1=CC=C(Cl)C=C1
Molecular Weight (g/mol) 
  • (3)
  • (3)
Percent Purity 
  • (3)
  • (3)
Quantity 
  • (2)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results