missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Ryuvidine
GREENER_CHOICE

Product Code. 18045372
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18045372 10 mg 10mg
18073972 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18045372 Supplier Tocris Bioscience™ Supplier No. 2609/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

SETD8 inhibitor; also CDK4 inhibitor

Inhibitor of SETD8 protein lysine methyltransferase (PKMT) (IC50 = 0.5 μM); suppresses H4K20 monomethylation in vitro. Also inhibits cyclin-dependent kinase (CDK) 4 (IC50 = 6.0 μM at CDK4/cyclin D1). Induces S phase accumulation in HEK293T cells. Cytotoxic against a range of human cancer cells.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 265312-55-8
Molecular Formula C15H12N2O2S

Specifications

Quantity 10 mg
Formula Weight 284.33
Purity >98%
Molecular Weight (g/mol) 284.333
Chemical Name or Material Ryuvidine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.