CAS RN 265312-55-8
IUPAC Name:
2-methyl-5-[(4-methylphenyl)amino]-4,7-dihydro-1,3-benzothiazole-4,7-dione
Synonyms:
2-methyl-5-[(4-methylphenyl)amino]-1,3-benzothiazole-4,7-dione
Molecular Weight (g/mol):
284.33
Molecular Formula:
C15H12N2O2S
InChi Key:
HFPLHASLIOXVGS-UHFFFAOYSA-N
SMILES:
CC1=NC2=C(S1)C(=O)C=C(NC1=CC=C(C)C=C1)C2=O