missing translation for 'onlineSavingsMsg'
Learn More

RY655 Rat Anti-Mouse IgM, BD OptiBuild™

Product Code. 32228444
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
32228444 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 32228444 Supplier BD Biosciences Supplier No. 774053
 Restricted Item
Estimated Shipment: 05-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Rat Monoclonal Secondary Antibody

Monoclonal Antibody Specifically detects in Mouse Samples. It is validated for Flow Cytometry.

The II/41 clone reacts specifically with mouse IgM of Igh-C[a] and Igh-C[b] haplotypes. It does not react with other Ig isotypes and has been reported not to stimulate B-cell proliferation. The BD Horizon RealYellow 655 (RY655) Dye is a tandem fluorochrome from the BD family of yellow-green dyes. It has an excitation maximum (Ex Max) at 557 nm and an emission maximum (Em Max) at 654 nm. RY655 can be used on both spectral and conventional cytometers and is designed to be excited by the Yellow-Green laser (561-nm). For conventional instruments equipped with a Yellow-Green laser (561-nm), RY655 can be used as an alternative to PE-Cy5 and an optical filter centered near 660 nm (e.g., a 660/30-nm or 670/30-nm bandpass filter) is recommended. For conventional and spectral instruments equipped with both a Blue (488-nm) and Yellow-Green (561-nm) laser and appropriate detectors, it can be used in conjunction with RB670.
TRUSTED_SUSTAINABILITY

Specifications

Applications Flow Cytometry
Classification Monoclonal
Clone II/41
Concentration 0.2 mg/mL
Conjugate RY655
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Gene Alias Igh-M; Igm; Immunoglobulin M heavy chain
Host Species Rat
Immunogen Not reported
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Secondary
Gene ID (Entrez) 16019
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions that minimize unconjugated dye and antibody.
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.