missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RUNX2/CBFA1 Antibody (CL0235) [DyLight 650], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-52888C
This item is not returnable.
View return policy
Description
RUNX2/CBFA1 Monoclonal antibody specifically detects RUNX2/CBFA1 in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),KnockDown,Immunocytochemistry/ Immunofluorescence
Specifications
| RUNX2/CBFA1 | |
| Monoclonal | |
| DyLight 650 | |
| Acute myeloid leukemia 3 protein, CBFA1, CBF-alpha-1, CCD1, CCDAML3, CLCD, Core-binding factor subunit alpha-1, core-binding factor, runt domain, alpha subunit 1, MGC120023, ML3, oncogene AML-3, OSF2, OSF-2, osteoblast-specific transcription factor 2, PEA2aA, PEA2-alpha A, PEBP2A, PEBP2aA, PEBP2-alpha A, polyomavirus enhancer-binding protein 2 alpha A subunit, runt domain, alpha subunit 1, runt related transcription factor 2, runt-related transcription factor 2, RUNX2, SL3/AKV core-binding factor alpha A subunit, SL3-3 enhancer factor 1 alpha A subunit | |
| This antibody was developed using a recombinant protein derived from Q13950, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Epigenetics, Growth and Development, Stem Cells, Transcription Factors and Regulators | |
| 860 | |
| Store at 4⪚C in the dark. | |
| IgG1 |
| Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), KnockDown, Immunocytochemistry/Immunofluorescence | |
| CL0235 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction