missing translation for 'onlineSavingsMsg'
Learn More

RUNX2/CBFA1 Antibody - BSA Free, Novus Biologicals™

Product Code. 18067583 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
Unit Size:
0.1mL
25µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18067583 0.025 mL 25µL
18756533 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18067583 Supplier Novus Biologicals Supplier No. NBP177461SS

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody has been used in 12 publications

RUNX2/CBFA1 Polyclonal specifically detects RUNX2/CBFA1 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RUNX2/CBFA1
Applications Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot reported in scientific literature (PMID 29208768), Immunohistochemistry 1:200, Immunocytochemistry/Immunofluorescence 1:100, Immunohistochemistry-Paraffin 1:200
Formulation PBS and 30% Glycerol with 0.05% Sodium Azide
Gene Accession No. Q13950
Gene Alias Acute myeloid leukemia 3 protein, AML3CCD, CBFA1MGC120022, Core-binding factor subunit alpha-1, MGC120023, PEA2-alpha A, PEBP2A, PEBP2A1CCD1, PEBP2aA1, PEBP2-alpha A, runt domain, alpha subunit 1, runt-related transcription factor 2, SL3/AKV core-binding factor alpha A subunit, SL3-3 enhancer factor 1 alpha A subunit
Gene Symbols RUNX2
Host Species Rabbit
Immunogen This RUNX2/CBFA1 Antibody was prepared from a synthetic peptide made to an internal region of the human RUNX2 protein (within residues 300-375). [Swiss-Prot Q13950]
Molecular Weight of Antigen 56.6 kDa
Purification Method Affinity Purified
Quantity 0.025 mL
Regulatory Status RUO
Research Discipline Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 860
Target Species Human, Mouse
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.