missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RUFY2 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-93691-0.1ml
This item is not returnable.
View return policy
Description
RUFY2 Polyclonal antibody specifically detects RUFY2 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| RUFY2 | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000 | |
| antigen MU-RMS-40.17, FLJ10063, KIAA1537, rab4-interacting protein related, RABIP4R, RUN and FYVE domain containing 2, RUN and FYVE domain-containing 2, RUN and FYVE domain-containing protein 2, Run- and FYVE-domain containing protein, ZFYVE13 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 144-403 of human RUFY2 (NP_001035882.1). IDFSMYLKNEEDIGNKERNVQIAAILDQKNYVEELNRQLNSTVSSLHSRVDSLEKSNTKLIEELAIAKNNIIKLQEENHQLRSENKLILMKTQQHLEVTKVDVETELQTYKHSRQGLDEMYNEARRQLRDESQLRQDVENELAVQVSMKHEIELAMKLLEKDIHEKQDTLIGLRQQLEEVKAINIEMYQKLQGSEDGLKEKNEIIARLEEKTNKITAAMRQLEQSDNDLLTQTRTIAMSLVKCASSDTQDQYKLVKDISF | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 55680 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction