missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RUFY1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
415.00€ - 631.00€
Specifications
| Antigen | RUFY1 |
|---|---|
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Target Species | Human |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18662890
|
Novus Biologicals
NBP2-76553-25ul |
25 μL |
415.00€
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18333301
|
Novus Biologicals
NBP2-76553 |
100 μL |
631.00€
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
RUFY1 Polyclonal specifically detects RUFY1 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
| RUFY1 | |
| Unconjugated | |
| Human | |
| La-binding protein 1, Rab4-interacting protein, RABIP4FYVE-finger protein EIP1, RUN and FYVE domain containing 1, RUN and FYVE domain-containing 1, RUN and FYVE domain-containing protein 1, ZFYVE12FLJ22251, Zinc finger FYVE domain-containing protein 12 | |
| RUFY1 | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Polyclonal | |
| Rabbit | |
| PBS, pH 7.2, containing 40% glycerol with 0.02% sodium azide | |
| 80230.0 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids: NLQMFHKAQNAESSLQQKNEAITSFEGKTNQVMSSMKQMEERLQHSERARQGAEERSHKLQQELGGRIGALQLQLSQLHEQCSSLEKEL | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title