Learn More
Eppendorf™ Rubber Mats
Product Code. 11893981 Shop All Eppendorf Products
missing translation for 'orderingAttributeHoverText'
For Use With (Equipment):
500mL rectangular bucket
Adapter 5702724009
Adapter 5702735000
Adapter 5702737003
Adapters for Rotor A-4-44
Rotor A-8-17
Rotor F-35-30-17
Capacity:
1 x 10 mL Vessel
1 x 15 mL Round Bottom Tube
1 x 15 mL Vessel
1 x 500 mL Bucket
1 x 7 mL Vessel
4 x 100 mL Tubes
missing translation for 'unitSize'
Each
Pack of 10
Pack of 16
Pack of 20
Pack of 4
Pack of 8
Product Code. 11893981
missing translation for 'mfr': Eppendorf™ 5702741000
For use with Eppendorf Adapters and buckets
  • Rubber mats
  • Available for a variety of different rotors and adapters
TRUSTED_SUSTAINABILITY
Specifications
Type Mat
Material Rubber
For Use With (Equipment) Adapter 5702737003
Color Black
Capacity 1 x 7 mL Vessel
Includes Set of 20 mats