missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ RTN4RL1 Monoclonal Antibody (319123)
GREENER_CHOICE

Mouse Monoclonal Antibody

Brand:  Invitrogen™ MA561224

Product Code. 30534852

  • 459.00€ / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Reconstitute at 0.5 mg/mL in sterile PBS.

RTN4RL1 may play a role in regulating axonal regeneration and plasticity in the adult central nervous system.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

RTN4RL1
Monoclonal
0.5 mg/mL
PBS with trehalose and no preservative
Q86UN2
Rtn4rl1
Mouse myeloma cell line NS0-derived recombinant human NgR3/NgRH2, Cys25-Ala419.
100 μg
Primary
Human
Antibody
IgG1
Western Blot
319123
Unconjugated
Rtn4rl1
NgR3; Ngrh2; NGRL2; Nogo receptor-like 2; Nogo-66 receptor homolog 2; Nogo-66 receptor-related protein 3; reticulon 4 receptor like 1; reticulon 4 receptor-like 1; reticulon-4 receptor-like 1; RGD:727812}; Rtn4rl1; rtn4rl1 {ECO:0000312
Mouse
Protein A/G
RUO
146760
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Lyophilized
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.