missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ (RS)-AMPA hydrobromide
GREENER_CHOICE

Product Code. 18084272
Change view
Click to view available options
Quantity:
1 mg
10 mg
50 mg
Unit Size:
10mg
1mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18084272 50 mg 50mg
18094812 1 mg 1mg
18075352 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 18084272 Supplier Tocris Bioscience™ Supplier No. 1074/50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Selective AMPA agonist; more water soluble form of (RS)-AMPA (Cat. No. 0169)

More water soluble hydrobromide salt of (RS)-AMPA (Cat. No. 0169).
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 171259-81-7
Molecular Formula C7H11BrN2O4

Specifications

Quantity 50 mg
Formula Weight 267.08
Purity >99%
Molecular Weight (g/mol) 267.079
Chemical Name or Material (RS)-AMPA hydrobromide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.