accessibility menu, dialog, popup

UserName

CAS RN 171259-81-7

CH3OHNOOOHH2NHBr

CAS RN 171259-81-7

IUPAC Name: 2-amino-3-(5-methyl-3-oxo-2,3-dihydro-1,2-oxazol-4-yl)propanoic acid hydrobromide
Synonyms: AMPA hydrobromide
Molecular Weight (g/mol): 267.08
Molecular Formula: C7H11BrN2O4
InChi Key: KUAHVIUZGLGASU-UHFFFAOYNA-N
SMILES: Br.CC1=C(CC(N)C(O)=O)C(=O)NO1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results