missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RRP12 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-93931-0.02ml
This item is not returnable.
View return policy
Description
RRP12 Polyclonal antibody specifically detects RRP12 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| RRP12 | |
| Polyclonal | |
| Western Blot 1:1000-1:2000 | |
| FLJ20231, KIAA0690DKFZp762P1116, ribosomal RNA processing 12 homolog (S. cerevisiae), RRP12-like protein | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-160 of human RRP12 (NP_055994.2). MGRSGKLPSGVSAKLKRWKKGHSSDSNPAICRHRQAARSRFFSRPSGRSDLTVDAVKLHNELQSGSLRLGKSEAPETPMEEEAELVLTEKSSGTFLSGLSDCTNVTFSKVQRFWESNSAAHKEICAVLAAVTEVIRSQGGKETETEYFAALMTTMEAVES | |
| 0.02 mL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 23223 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction