missing translation for 'onlineSavingsMsg'
Learn More

rRNA Antibody (Y10b) - BSA Free, Novus Biologicals™

Product Code. 18795153 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
Unit Size:
0.1mL
25µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18795153 0.025 mL 25µL
18776183 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18795153 Supplier Novus Biologicals Supplier No. NB100662SS

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody has been used in 21 publication

rRNA Monoclonal specifically detects rRNA in Human, Mouse, All Species, Bacteria samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Flow (Intracellular), Immunocytochemistry, CyTOF-reported, CyTOF-ready.
TRUSTED_SUSTAINABILITY

Specifications

Antigen rRNA
Applications Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), CyTOF
Classification Monoclonal
Clone Y10b
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Flow Cytometry 2.5 ug/ml, Immunohistochemistry 1:100 - 1:300, Immunocytochemistry/Immunofluorescence reported in scientific literature (PMID 34980897), Immunoprecipitation 10 ul/lysate, Immunohistochemistry-Paraffin 1:100 - 1:300, Flow (Intracellular) 2.5 ug/ml, Immunocytochemistry 1 - 3 ug/ml, CyTOF-reported, CyTOF-ready reported by customer review
Gene Alias ribosomal RNA 5.8s
Host Species Mouse
Immunogen The whole 5.8S ribosomal RNA.
Purification Method Protein A purified
Quantity 0.025 mL
Regulatory Status RUO
Research Discipline DNA Repair
Primary or Secondary Primary
Test Specificity This is specific for ribosomal RNA. There is no cross-reactivity with other RNAs.
Target Species Human, Mouse, All species, Bacteria
Content And Storage Store at 4C. Do not freeze.
Form Purified
Isotype IgG3 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.