missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RRM1 Rabbit anti-Human, Mouse, Non-human primate, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10526-1-AP-20UL
This item is not returnable.
View return policy
Description
RRM1 (Ribonucleotide reductase M1 polypeptide) is one of two non-identical subunits that constitute ribonucleoside-diphosphate reductase, an enzyme which catalyzes the biosynthesis of deoxyribonucleotides from the corresponding ribonucleotides. It provides the precursors necessary for DNA synthesis. RRM1 is involved in carcinogenesis, tumor progression.Specifications
| RRM1 | |
| Polyclonal | |
| Unconjugated | |
| RRM1 | |
| R1, ribonucleotide reductase M1, RIR1, RR1, RRM1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 20133, 6240, 685579 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Immunofluorescence, Immunoprecipitation, Immunocytochemistry, Immunohistochemistry (Paraffin) | |
| 0.22 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P07742, P23921 | |
| RRM1 | |
| RRM1 Fusion Protein Ag0789 | |
| 20 μL | |
| Primary | |
| Rat, Non-human Primate, Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction