missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RPS19BP1 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-93262-0.1ml
This item is not returnable.
View return policy
Description
RPS19BP1 Polyclonal antibody specifically detects RPS19BP1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
Specifications
| RPS19BP1 | |
| Polyclonal | |
| Western Blot 1:500-1:2000, Immunocytochemistry/ Immunofluorescence 1:50-1:200 | |
| 40S ribosomal protein S19-binding protein 1, AROS, dJ1104E15.4, FLJ21770, homolog of mouse S19 binding protein, MGC52010RPS19-binding protein 1, ribosomal protein S19 binding protein 1, S19BPactive regulator of SIRT1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-136 of human RPS19BP1 (NP_919307.1). MSAALLRRGLELLAASEAPRDPPGQAKPRGAPVKRPRKTKAIQAQKLRNSAKGKVPKSALDEYRKRECRDHLRVNLKFLTRTRSTVAESVSQQILRQNRGRKACDRPVAKTKKKKAEGTVFTEEDFQKFQQEYFGS | |
| 0.1 mL | |
| Cell Biology | |
| 91582 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction