missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RPRM Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
198.00€ - 468.00€
Specifications
| Antigen | RPRM |
|---|---|
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
RPRM Polyclonal antibody specifically detects RPRM in Mouse samples. It is validated for Western BlotSpecifications
| RPRM | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Cell Cycle and Replication | |
| PBS (pH 7.3), 50% glycerol | |
| 56475 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Mouse | |
| candidate mediator of the p53 dependent G2 arrest, candidate mediator of the p53-dependent G2 arrest, FLJ90327, protein reprimo, REPRIMO, reprimo, TP53 dependant G2 arrest mediator candidate, reprimo, TP53 dependent G2 arrest mediator candidate | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-109 of human RPRM (NP_062819.1). MNPALGNQTDVAGLFLANSSEALERAVRCCTQASVVTDDGFAEGGPDERSLYIMRVVQIAVMCVLSLTVVFGIFFLGCNLLIKSEGMINFLVKDRRPSKEVEAVVVGPY | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title