missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RPRD1B Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
183.00€ - 322.00€
Specifications
| Antigen | RPRD1B |
|---|---|
| Dilution | Western Blot 1:200-1:500, ELISA 1:5000-1:10000, Immunocytochemistry/ Immunofluorescence 1:50-1:200, Immunoprecipitation 1-5ul/mg of lysate |
| Applications | Western Blot, ELISA, Immunofluorescence, Immunoprecipitation |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18660177
|
Novus Biologicals
NBP3-12539-100ul |
100 μg |
322.00€
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18681867
|
Novus Biologicals
NBP3-12539-50ul |
50 μg |
183.00€
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
RPRD1B Polyclonal antibody specifically detects RPRD1B in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence, ImmunoprecipitationSpecifications
| RPRD1B | |
| Western Blot, ELISA, Immunofluorescence, Immunoprecipitation | |
| Unconjugated | |
| Rabbit | |
| Human | |
| Cell cycle-related and expression-elevated protein in tumor, cell-cycle related and expression-elevated protein in tumor, chromosome 20 open reading frame 77, CREPTC20orf77, dJ1057B20.2, DKFZp434P0735, FLJ44520, NET60, regulation of nuclear pre-mRNA domain containing 1B, regulation of nuclear pre-mRNA domain-containing protein 1B | |
| Produced in rabbits immunized with purified, recombinant Human RPRD1B (Accession#: AAH33629.1; Met1-Asp326) | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot 1:200-1:500, ELISA 1:5000-1:10000, Immunocytochemistry/ Immunofluorescence 1:50-1:200, Immunoprecipitation 1-5ul/mg of lysate | |
| Polyclonal | |
| Purified | |
| RUO | |
| 0.2 um filtered solution in PBS | |
| 58490 | |
| IgG | |
| Antigen and protein A Affinity-purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title