missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RPP20 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-94597-0.02ml
This item is not returnable.
View return policy
Description
RPP20 Polyclonal antibody specifically detects RPP20 in Human, Rat samples. It is validated for Western Blot
Specifications
| RPP20 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| EC 3.1.26.5, hPop5, HSPC004, processing of precursor 5, ribonuclease P/MRP subunit (S. cerevisiae), ribonuclease P/MRP protein subunit POP5, RPP2, RPP20 | |
| A synthetic peptide corresponding to a sequence within amino acids 50-150 of human POP5 (NP_057002.2). SIGFAVRYLNAYTGIVLLRCRKEFYQLVWSALPFITYLENKGHRYPCFFNTLHVGGTIRTCQKFLIQYNRRQLLILLQNCTDEGEREAIQKSVTRSCLLEE | |
| 0.02 mL | |
| Endocrinology | |
| 51367 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Rat | |
| Purified |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu