missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RPLP1 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-94116-0.1ml
This item is not returnable.
View return policy
Description
RPLP1 Polyclonal antibody specifically detects RPLP1 in Human, Mouse, Rat, Zebrafish samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
Specifications
| RPLP1 | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, Immunocytochemistry/ Immunofluorescence 1:50 - 1:100 | |
| acidic ribosomal phosphoprotein P1, FLJ27448, MGC5215, P1,60S acidic ribosomal protein P1, ribosomal protein, large, P1, RPP1, RRP1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-114 of human RPLP1 (NP_000994.1). MASVSELACIYSALILHDDEVTVTEDKINALIKAAGVNVEPFWPGLFAKALANVNIGSLICNVGAGGPAPAAGAAPAGGPAPSTAAAPAEEKKVEAKKEESEESDDDMGFGLFD | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat, Zebrafish | |
| Purified |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 6176 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction