missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RPL5 Rabbit Polyclonal Antibody, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 29092-1-AP-20UL
This item is not returnable.
View return policy
Description
The RPL5 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human RPL5. This antibody recognizes human, monkey antigen. The RPL5 antibody has been validated for the following applications: WB, IHC, ELISA analysis.Specifications
| RPL5 | |
| Polyclonal | |
| Unconjugated | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 6125 | |
| Store at -20°C. | |
| Polyclonal Antibody | |
| IgG |
| Western Blot, Immunohistochemistry, ELISA | |
| 500 μg/mL | |
| DBA6; MGC117339; MSTP030 | |
| Recombinant Protein | |
| 20 μL | |
| Primary | |
| Human, Monkey | |
| 34 kDa | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction