missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NF2/Merlin Antibody (S07-1A6), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-19715-50ul
This item is not returnable.
View return policy
Description
NF2/Merlin Monoclonal antibody specifically detects NF2/Merlin in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| NF2/Merlin | |
| Monoclonal | |
| 0.3 mg/mL | |
| 50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot | |
| S07-1A6 | |
| Unconjugated | |
| ACN, BANF, merlin, moesin-ezrin-radixin like, Moesin-ezrin-radixin-like protein, moesin-ezrin-radizin-like protein, neurofibromin 2 (bilateral acoustic neuroma), neurofibromin 2 (merlin), neurofibromin-2, SCH, Schwannomerlin, Schwannomin | |
| A synthetic peptide of human NF2/Merlin (Uniprot # P35240) | |
| 50 μg | |
| Cancer, Cell Cycle and Replication, Phospho Specific, Tumor Suppressors | |
| 4771 | |
| Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction