missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RPL18A Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14653-1-AP-150UL
This item is not returnable.
View return policy
Description
This gene encodes a protein belonging to the pantothenate kinase family. Pantothenate kinase is a key regulatory enzyme in the biosynthesis of coenzyme A (CoA) in bacteria and mammalian cells. It catalyzes the first committed step in the universal biosynthetic pathway leading to CoA and is itself subject to regulation through feedback inhibition by CoA. This family member is most abundant in muscle but is expressed in all tissues.Specifications
| RPL18A | |
| Polyclonal | |
| Unconjugated | |
| RPL18A | |
| 60S ribosomal protein L18a, ribosomal protein L18a, RPL18A | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 290641, 6142, 76808 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| 0.23 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P62717, P62718, Q02543 | |
| Rpl18a | |
| RPL18A Fusion Protein Ag6251 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction