missing translation for 'onlineSavingsMsg'
Learn More

RPA2 Mouse Monoclonal Antibody, Proteintech

Mouse Monoclonal Antibody

Brand:  Proteintech 67999-1-IG-150UL

 View more versions of this product

Product Code. 30233675

  • ONLINE EXCLUSIVE PRICE
    410.00€
    349.00€ / 150µL
    Save 61.00€
    14% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

The RPA2 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human RPA2. This antibody recognizes human, mouse, rat, pig, rabbit antigen. The RPA2 antibody has been validated for the following applications: WB, IF/ICC, ELISA analysis.
Specifications

Specifications

RPA2
Monoclonal
1000 μg/mL
REPA2; RPA32
Recombinant Protein
150 μL
Primary
Human, Mouse, Rat, Pig, Rabbit
30 kDa
Liquid
Western Blot, Immunocytochemistry/Immunofluorescence, ELISA
1B9H11
Unconjugated
Mouse
Protein G purification
RUO
6118
Store at -20°C. Stable for one year after shipment.
Monoclonal Antibody
IgG1
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.