missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RP2 Mouse Monoclonal Antibody, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67795-1-IG-150UL
This item is not returnable.
View return policy
Description
The RP2 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human RP2. This antibody recognizes human, rabbit antigen. The RP2 antibody has been validated for the following applications: WB, ELISA analysis.Specifications
| RP2 | |
| Monoclonal | |
| 1000 μg/mL | |
| DELXp11.3; KIAA0215; TBCCD2 | |
| Recombinant Protein | |
| 150 μL | |
| Primary | |
| Human, Rabbit | |
| 37-40 kDa | |
| Liquid |
| Western Blot, ELISA | |
| 1G2A9 | |
| Unconjugated | |
| Mouse | |
| Protein G purification | |
| RUO | |
| 6102 | |
| Store at -20°C. | |
| Monoclonal Antibody | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction