missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- Fixed-angle rotor R9A2
- Includes 4 x 1500 polypropylene bottle (WM)
- For centrifuge CR22N
- Max. capacity: 4 x 1,500 mL (nominal capacity)
- Max. speed: 15,100 x g (8,500 rpm)
- For harvesting of bacteria, cells or yeast for e.g. recombinant protein or antibody purification in easy-to-handle, aerosol-tight bottles with 4 x 1.5 L
- Made from high-quality aluminum
Specifications
Specifications
| Material | Aluminum |
| Type | Fixed Angle Rotor |
| Capacity | 4 x 1500 mL |
| Max. RCF | 15,100 x g |
| Max. Speed | 8,500 rpm |
| For Use With (Equipment) | Centrifuge CR22N/CR21N |
| Biocontainment | No |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?