missing translation for 'onlineSavingsMsg'
Learn More

Eppendorf Rotor R8S for Centrifuge CR22N

Product Code. 31945128 Shop All Eppendorf Products
Change view
Click to view available options
Capacity:
4 x 50 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity Unit Size
31945128 4 x 50 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 31945128 Supplier Eppendorf Supplier No. 5721202400
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Swing-bucket rotor is designed for the strong pelleting of bacteria, yeast or cells for e.g. recombinant protein purification in conical tubes with 4 x 50 mL.

  • Swing-bucket rotor R8S for strong pelleting of bacteria, yeast or cells for e.g. recombinant protein purification in conical tubes
  • Includes 4 buckets for 15/50 mL Eppendorf conical tubes
  • For centrifuge CR22N
  • Max. capacity: 4 x 50mL (nominal capacity)
  • Max. speed: 11,500 x g (8,000 rpm)
  • Made from high-quality aluminum
  • No consumables included
TRUSTED_SUSTAINABILITY

Specifications

Material Aluminum
Type Swinging Bucket Rotor
Capacity 4 x 50 mL
Max. RCF 11,500 x g
Max. Speed 8,000 rpm
For Use With (Equipment) Centrifuge CR22N
Biocontainment No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.