Learn More
Eppendorf Rotor R5S4 for Centrifuge CR30NX
Product Code. 31945217 Shop All Eppendorf Products
Change view
Click to view available options
Max. RCF:
3,100 x g
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Max. RCF Unit Size
31945217 3,100 x g Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 31945217 Supplier Eppendorf Supplier No. 5721302401
 Restricted Item
Estimated Shipment: 06-10-2026
Add to basket
Add to basket
Swing-bucket rotor compatible with the CR30NX centrifuge.
  • Swing-bucket rotor R5S4
  • Aerosol-tight,
  • For up to 12 microplates
  • For centrifuge CR30NX
  • Max. speed: 3,100 x g (4,700 rpm)
  • For plate centrifugation for e.g. ELISA or PCR in up to 12 x 96 plates
  • Made from high-quality aluminum
TRUSTED_SUSTAINABILITY
Specifications
Material Aluminum
Type Swinging Bucket Rotor
Max. RCF 3,100 x g
Max. Speed 4,700 rpm
For Use With (Equipment) Centrifuge CR30NX
Biocontainment No