missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- Swing-bucket rotor R5S4
- For up to 12 microplates
- Aerosol-tight
- For centrifuge CR22N
- Max. speed: 3,100 x g (4,700 rpm)
- For plate centrifugation for e.g. ELISA or PCR in up to 12 x 96 plates
- Made from high-quality aluminum
- K-factor: 11,761
- Borehole angle: 90°
- No consumables included
Specifications
Specifications
| Material | Aluminum |
| Type | Swinging Bucket Rotor |
| Max. RCF | 3,100 x g |
| Max. Speed | 4,700 rpm |
| K Factor | 11,761 |
| For Use With (Equipment) | Centrifuge CR22N |
| Biocontainment | No |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?