Learn More
Eppendorf™ Rotor for Centrifuges 5910 Ri and 5920 R
Shop All Eppendorf Products
Click to view available options
Capacity:
20 × 5 mL
4 to 750 mL
4 x 1,000 mL
4 x 750 mL
48 x 2 mL
6 x 250 mL
6 × 50 mL
Max. Speed:
10,100 rpm
13,100 rpm
13,500 rpm
13,700 rpm
4,500 rpm
4,700 rpm
4500 rpm
Max. RCF:
15,054 x g
20,130 x g
20,913 x g
21,194 x g
4,347 x g
4,800 x g
Includes:
Eppendorf QuickLock™ Rotor Lid
Rotor Lid
Round Buckets
Universal Buckets
Without Buckets
missing translation for 'unitSize'
Each
Description
- Precision-engineered for optimal sample separation.
- All fixed-angle rotors, rotor lids, buckets, caps and adapters can be autoclaved (121°C, 20 min.).
- Accessories (adaptors and buckets) further customize your centrifuge setup.
- Aerosol-tight options available.
Specifications
Specifications
| Material | Anodized Aluminum |
| Type | Rotor |
| Capacity | 20 × 5 mL |
| Max. RCF | 20,913 x g |
| Max. Speed | 13,100 rpm |
| For Use With (Equipment) | Centrifuges 5910 Ri and 5920 R |
| Includes | Eppendorf QuickLock™ Rotor Lid |
| Model | FA-20x5, Aerosol-tight |