Learn More
Eppendorf™ Rotor Adapters for Centrifuge 5427 R
For 1.5/2mL tubes and cryotubes
42.75€
Specifications
Type | Rotor Adapter |
---|---|
For Use With (Equipment) | Rotors FA-45-12-17, FA-45-16-17 and FA-45-20-17 |
No. per Pack | Set of 4 |
Product Code | Brand | Capacity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity | Price | Quantity & Availability | |||||
13216619
|
Eppendorf™
5820768002 |
1 x 1.5/2 mL Vessel |
42.75€
Pack of 10 |
Estimated Shipment: 28-05-2024
Log in to see stock. |
|||||
13226619
|
Eppendorf™
5820769009 |
1 Cryotube |
42.75€
Pack of 4 |
Estimated Shipment: 28-05-2024
Log in to see stock. |
|||||
Description
- Rotors FA-45-12-17, FA-45-16-17 and FA-45-20-17
Specifications
Rotor Adapter | |
Rotors FA-45-12-17, FA-45-16-17 and FA-45-20-17 | |
Set of 4 |