Learn More
Thermo Scientific Chemicals Rotigotine
Product Code. p-7214736 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
Product Code. 17281242
Brand: Thermo Scientific Chemicals 466541000
Request Bulk or Custom Format
CAS: 99755-59-6 | C19H25NOS, C19H25NOS | 315.48 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 99755-59-6
Molecular Formula C19H25NOS
Specifications
Quantity 100 mg
Formula Weight 315.47
InChI Key KFQYTPMOWPVWEJ-UHFFFAOYNA-N
SMILES CCCN(CCC1=CC=CS1)C1CCC2=C(O)C=CC=C2C1
IUPAC Name 6-{propyl[2-(thiophen-2-yl)ethyl]amino}-5,6,7,8-tetrahydronaphthalen-1-ol
Molecular Weight (g/mol) 315.48
Chemical Name or Material Rotigotine