accessibility menu, dialog, popup

UserName

CAS RN 99755-59-6

H3CNSHO

CAS RN 99755-59-6

IUPAC Name: 6-{propyl[2-(thiophen-2-yl)ethyl]amino}-5,6,7,8-tetrahydronaphthalen-1-ol
Synonyms: rotigotine transdermal patch rotigotine
Molecular Weight (g/mol): 315.48
Molecular Formula: C19H25NOS
InChi Key: KFQYTPMOWPVWEJ-UHFFFAOYNA-N
SMILES: CCCN(CCC1=CC=CS1)C1CCC2=C(O)C=CC=C2C1
Molecular Weight (g/mol) 
  • (2)
  • (2)
Certifications/Compliance 
  • (1)
  • (1)
Industry Type 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results