missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ROR2 Mouse anti-Human, Clone: ROR2/1912, Novus Biologicals™
Mouse Monoclonal Antibody
484.00€
Specifications
| Antigen | ROR2 |
|---|---|
| Clone | ROR2/1912 |
| Concentration | 0.2 mg/mL |
| Dilution | Immunohistochemistry-Paraffin 1 to 2 μg/mL |
| Applications | Immunohistochemistry (Paraffin) |
Description
ROR2 Monoclonal specifically detects ROR2 in Human samples. It is validated for Immunohistochemistry-Paraffin.Specifications
| ROR2 | |
| 0.2 mg/mL | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Protein Kinase, Wnt Signaling Pathway | |
| 10 mM PBS with 0.05% BSA | |
| 4920 | |
| IgG | |
| Protein A or G purified |
| ROR2/1912 | |
| Immunohistochemistry-Paraffin 1 to 2 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| BDB, BDB1, EC 2.7.10.1, MGC163394, neurotrophic tyrosine kinase receptor-related 2, Neurotrophic tyrosine kinase, receptor-related 2, NTRKR2tyrosine-protein kinase transmembrane receptor ROR2, receptor tyrosine kinase-like orphan receptor 2 | |
| Human cultured keratinocytes | |
| Primary | |
| Store at 4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title