Learn More
Novus Biologicals™ ROBO1 Antibody (Asclepius Technology patent anti-Robo1 CAR) - Chimeric, Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-28772-1mg
This item is not returnable.
View return policy
Description
ROBO1 Monoclonal antibody specifically detects ROBO1 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications
| ROBO1 | |
| Monoclonal | |
| Unconjugated | |
| Q9Y6N7 | |
| Human | |
| Protein A purified | |
| RUO | |
| Primary | |
| Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex. | |
| Store at -20°C in powder form. Store at -80°C once reconstituted. | |
| IgG1 |
| Flow Cytometry, ELISA, Functional Assay | |
| Asclepius Technology patent anti-Robo1 CAR | |
| Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2) | |
| DUTT1Drosophila) homolog 1, H-Robo-1, MGC131599, MGC133277, roundabout, axon guidance receptor, homolog 1 (Drosophila) | |
| ROBO1 | |
| 1 mg | |
| Angiogenesis, Cancer, Extracellular Matrix | |
| 6091 | |
| Human | |
| Purified |