missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Ro 67-7476
Description
Positive allosteric modulator of mGlu1 receptors. Potentiates glutamate-induced calcium release (EC50 = 60.1 nM at rat mGluR1a). Displays no activity at human mGlu1 receptors.
Specifications
Specifications
| Quantity | 10 mg |
| Product Line | Tocris |
| Formula Weight | 319.39 |
| Purity | >99% |
| Molecular Weight (g/mol) | 319.394 |
| Chemical Name or Material | Ro 67-7476 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?