accessibility menu, dialog, popup

UserName

CAS RN 298690-60-5

CH3SOONF

CAS RN 298690-60-5

IUPAC Name: 2-(4-fluorophenyl)-1-(4-methylbenzenesulfonyl)pyrrolidine
Synonyms: 2-(4-fluorophenyl)-1-(4-methylbenzenesulfonyl)pyrrolidine
Molecular Weight (g/mol): 319.39
Molecular Formula: C17H18FNO2S
InChi Key: DAEHFYNGSSBGSS-UHFFFAOYNA-N
SMILES: CC1=CC=C(C=C1)S(=O)(=O)N1CCCC1C1=CC=C(F)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results