missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RNPEPL1 Antibody (11G1), Novus Biologicals™
Mouse Monoclonal Antibody
187.00€
Specifications
| Antigen | RNPEPL1 |
|---|---|
| Clone | 11G1 |
| Dilution | Western Blot 1:100 - 1:2000 |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
Description
RNPEPL1 Monoclonal specifically detects RNPEPL1 in Non-species specific samples. It is validated for Western Blot.Specifications
| RNPEPL1 | |
| Western Blot 1:100 - 1:2000 | |
| Unconjugated | |
| Mouse | |
| argininyl aminopeptidase-like 1, arginyl aminopeptidase (aminopeptidase B)-like 1, arginyl aminopeptidase-like 1, EC 3.4.11.-, FLJ10806, FLJ26675, MGC99544, RNPEP-like 1 | |
| RNPEPL1 | |
| IgG3 | |
| Protein G purified |
| 11G1 | |
| Monoclonal | |
| Purified | |
| RUO | |
| 57140 | |
| Synthetic N-terminal arginylated peptide conjugated to KLH | |
| Primary | |
| Specific for N-terminal arginine, next to both glutamic acid and aspartic acid. Does not detect internal arginine. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title