missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RNASEH2A Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 2 publications
189.00€
Specifications
| Antigen | RNASEH2A |
|---|---|
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1 μg/mL, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 2-4 μg/mL, Immunoprecipitation |
| Applications | Western Blot |
| Classification | Polyclonal |
Description
RNASEH2A Polyclonal specifically detects RNASEH2A in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunoprecipitation, Immunofluorescence, Immunocytochemistry.Specifications
| RNASEH2A | |
| Western Blot 1 μg/mL, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 2-4 μg/mL, Immunoprecipitation | |
| Polyclonal | |
| Rabbit | |
| DNA replication Transcription Translation and Splicing | |
| PBS with 0.02% Sodium Azide | |
| AGS4RNase H2 subunit A, Aicardi-Goutieres syndrome 4, Aicardi-Goutieres syndrome 4 protein, JUNB, ribonuclease H2 subunit A, ribonuclease H2, large subunit, ribonuclease H2, subunit A, Ribonuclease HI large subunit, Ribonuclease HI subunit A, ribonuclease HI, large subunit, RNase H(35), RNASEHIEC 3.1.26.4, RNHIARNase HI large subunit, RNHL | |
| RNASEH2A | |
| IgG | |
| Affinity Purified | |
| 33 kDa |
| 1 mg/mL | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Human | |
| O75792 | |
| 10535 | |
| Antibody was raised against a 17 amino acid synthetic peptide near the center of human RNAse H2A. The immunogen is located within amino acids 190 - 240 of RNAse H2A. Amino Acid Squence: LDTDYGSGYPNDPKTKA | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title