Learn More
RNase T2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13753-1-AP-20UL
This item is not returnable.
View return policy
Description
RNASET2 is a novel member of the Rh/T2/S-glycoprotein class of extracellular ribonucleases. It is a single copy gene that maps to 6q27, a region associated with human maligcies and chromosomal rearrangement, and has been suggested to function as a tumor suppressor protein. Its expression is suppressed in Human T-cell Leukemia Virus type 1 (HTLV-1) infected cells following the binding of the HTLV-1 Tax protein to the RNASET2 promoter. As Adult T-cell leukemia (ATL) is one of the primary diseases caused by HTLV-1 infection, a reduction in the level of RNASET2 by Tax may play a role in ATL development.Specifications
| RNase T2 | |
| Polyclonal | |
| Unconjugated | |
| RNASET2 | |
| Ribonuclease 6, Ribonuclease T2, RNASE6PL, RNASET2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 100037283, 8635 | |
| -20°C | |
| Liquid |
| Immunofluorescence, Immunoprecipitation, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.13 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| C0HKG5, O00584 | |
| RNASET2, Rnaset2a | |
| RNase T2 Fusion Protein Ag4695 | |
| 20 μL | |
| Primary | |
| Mouse, Human | |
| Antibody | |
| IgG |