missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RNA Polymerase II/POLR2A Antibody (POLR2A/9089R), Novus Biologicals™
Rabbit Monoclonal Antibody
268.00€ - 496.00€
Specifications
| Antigen | RNA Polymerase II/POLR2A |
|---|---|
| Clone | POLR2A/9089R |
| Concentration | 0.2 mg/mL |
| Dilution | Western Blot : 1-2 μg/mL, Immunohistochemistry-Paraffin : 1-2 μg/mL |
| Applications | Western Blot, Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18661285
|
Novus Biologicals
NBP3-23600-100ug |
100 μg |
496.00€
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18668644
|
Novus Biologicals
NBP3-23600-20ug |
20 μg |
268.00€
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
RNA Polymerase II/POLR2A Monoclonal antibody specifically detects RNA Polymerase II/POLR2A in Human, Yeast samples. It is validated for Western Blot, Immunohistochemistry (Paraffin)Specifications
| RNA Polymerase II/POLR2A | |
| 0.2 mg/mL | |
| Western Blot, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Chromatin Research, Epigenetics, Phospho Specific, Transcription Factors and Regulators | |
| 10 mM PBS with 0.05% BSA | |
| 5430 | |
| IgG κ | |
| Protein A purified |
| POLR2A/9089R | |
| Western Blot : 1-2 μg/mL, Immunohistochemistry-Paraffin : 1-2 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Yeast | |
| DNA-directed RNA polymerase II largest subunit, RNA polymerase II 220 kdsubunit, DNA-directed RNA polymerase II subunit A, DNA-directed RNA polymerase II subunit RPB1, DNA-directed RNA polymerase III largest subunit, EC 2.7.7.48, EC 2.7.7.6, hRPB220, hsRPB1, MGC75453, POLR2, POLRA, polymerase (RNA) II (DNA directed) polypeptide A (220kD), polymerase (RNA) II (DNA directed) polypeptide A, 220kDa, RNA Pol 2, RNA Polymerase 2, RNA polymerase II subunit B1, RNA-directed RNA polymerase II subunit RPB1, RNAPII, RPB1, RPBh1, RpIILS, RPO2, RPOL2 | |
| Ten repeats of synthetic peptide YSPTSPS using chemically synthesized phospho-Ser5 | |
| Primary | |
| Store at 4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title