missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ MDP1 Protein
Shop All Bio Techne Products
Click to view available options
Quantity:
0.1 mg
0.5 mg
Unit Size:
0.1mg
0.5mg
Description
A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-176 of Human MDP1 The Recombinant Human MDP1 Protein is derived from E. coli. The Recombinant Human MDP1 Protein has been validated for the following applications: SDS-Page.
Specifications
Specifications
| Concentration | 0.5mg/mL |
| For Use With (Application) | SDS-PAGE |
| Formulation | Liquid. 20mM Tris-HCl buffer (pH 8.0) containing 10% glycerol, 2mM DTT, 100mM NaCl |
| Molecular Weight (g/mol) | 22.7kDa |
| Purification Method | SDS-PAGE |
| Quantity | 0.1 mg |
| Storage Requirements | Store at -80°C. Avoid freeze-thaw cycles. |
| Regulatory Status | RUO |
| Common Name | MDP1 |
| Conjugate | Unconjugated |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur